missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CHRDL1 Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
Description
CHRDL1 is an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
| Accession Number | Q9BU40 |
| For Use With (Application) | Functional Assay |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 91851 |
| Molecular Weight (g/mol) | 48.8 kDa |
| Name | CHRDL1 |
| Quantity | 10 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction