Learn More
R&D Systems™ Human Connexin 50/GJA8 (NP_005258) VersaClone cDNA
Product Code. 18775068
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18775068 10 μg 10µg
1 options
Product Code. 18775068 Supplier R&D Systems™ Supplier No. RDC0927
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 10µg
    Save 18.65 €
    5% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Gap junction protein, alpha 8, 50kDa
The Human Connexin 50/GJA8 (NP_005258) VersaClone cDNA clone contains the complete ORF for Connexin 50/GJA8, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Connexin 50/GJA8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans