missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CXCL10 (IP-10) Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
50µg
5µg
Description
Recombinant Human IP-10 is an 8.6 kDa protein consisting of 77 amino acids, including the four conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
Specifications
Specifications
| Accession Number | P02778 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3627 |
| Molecular Weight (g/mol) | 8.6 kDa |
| Name | Human CXCL10 (IP-10) |
| Quantity | 25 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction