missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Cytochrome c Quantikine ELISA Kit
Description
The Human Cytochrome c Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Cytochrome c in cell lysates (20 ul), subcellular fractions (100 ul).
Specifications
Specifications
| Assay Range | 0.63 - 20ng/mL |
| Assay Sensitivity | 0.31ng/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Product Line | R&D Systems |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Lysates, Subcellular Fractions |
| Specificity | Natural Human Cytochrome C |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?