missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human DAP12 (NP_003323) VersaClone cDNA
Description
- TYROBP TYRO protein tyrosine kinase binding protein [ Homo sapiens (human) ]
- Also known as: DAP12; KARAP; PLOSL
- The DAP12 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- DAP12 is a transmembrane signaling polypeptide which contains an immunoreceptor tyrosine-based activation motif (ITAM) in its cytoplasmic domain.
- It may associate with the killer-cell inhibitory receptor (KIR) family of membrane glycoproteins and may act as an activating signal transduction element.
- DAP12 may bind zeta-chain (TCR) associated protein kinase 70kDa (ZAP-70) and spleen tyrosine kinase (SYK) and play a role in signal transduction, bone modeling, brain myelination, and inflammation.
- Alternatively spliced transcripts encoding different proteins have been described.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hTYROBP |
| Accession Number | NP_003323 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 355 bp |
| Gene ID (Entrez) | 7305 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?