missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNMT3A (aa 86-100) Synthetic Peptide

Product Code. 11534770
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11534770 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11534770 Supplier Invitrogen™ Supplier No. PEP116

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Human Dnmt3a (residues 86-100) Synthetic Peptide, PEP-116, from Invitrogen.

Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, 3 families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. Dnmt2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. The Dnmt3 family members, Dnmt3a and Dnmt3b, are strongly expressed in ES cells but their expression is down regulated in differentiating ES cells and is low in adult somatic tissue. Recently, it has been shown that naturally occurring mutations of Dnmt3b gene occurs in patients with a rare autosomal recessive disorder, termed ICF (immunodeficiency, centromeric instability, and facial anomalies) syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 1788
Species Human
Purification Method purified
Accession Number Q9Y6K1
Common Name Human DNMT3A (aa 86-100)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol DNMT3A
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence D(86)-L-L-P-N-G-D-L-E-K-R-S-E-P-Q(100)-C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.