missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DPP4 (967 amino acids) native Protein from human serum

Product Code. 16098076
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16098076 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16098076 Supplier Abnova™ Supplier No. P4922.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for Func, SDS-PAGE

The protein encoded by this gene is identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. [provided by RefSeq]

Specifications

For Use With (Application) Functional Study, SDS-PAGE
Formulation In 20mM Tris-HCl, 500mM NaCl, 1mM CaCl
Gene ID (Entrez) 1803
Molecular Weight (g/mol) 105kDa
Name DPP4 (Human) Native Protein
Preparation Method Native protein purified from human serum
Purification Method Saltprecipitation/Ion Exchange Chromatography/Gelfiltration
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ADABP/ADCP2/CD26/DPPIV/TP103
Common Name DPP4
Gene Symbol DPP4
Biological Activity > 200mU/mg
Species Human
Protein Tag None
Expression System Native protein purified from human serum
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.