missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human DPPIV / CD26 Protein, His Tag (Active dimer) (active enzyme)

Product Code. 15886587
Change view
Click to view available options
Quantity:
200 μg
50 μg
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15886587 50 μg 50µg
15876587 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15886587 Supplier ACROBiosystems Supplier No. DP4H522150UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dipeptidyl peptidase-IV (DPPIV) is also known as adenosine deaminase complexing protein 2, DPPIV or CD26 is antigenic enzyme expressed on the surface of most cell types and is associated with immune regulation, signal transduction and apoptosis. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. The substrates of DPPIV are proline (or alanine)-containing peptides and include growth factors, chemokines, neuropeptides, and vasoactive peptides. DPPIV plays a major role in glucose metabolism. It is responsible for the degradation of incretins such as GLP-1. DPPIV plays an important role in tumor biology, and is useful as a marker for various cancers, with its levels either on the cell surface or in the serum increased in some neoplasms and decreased in others. DPPIV also binds the enzyme adenosine deaminase specifically and with high affinity. The significance of this interaction has yet to be established.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.7 kDa. The protein migrates as 95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_001926.2
Molecular Weight (g/mol) 85.7 kDa
Quantity 50 μg
Source Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 34 - Pro 766 (Accession # NP_001926.2).
Immunogen Asp 34 - Pro 766
Storage Requirements -20°C
Gene Alias DPP4,ADABP,ADCP2,CD26,DPPIV,TP103
Common Name DPPIV
Conjugate Unconjugated
Species Human
Protein Tag His Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Enzymes
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.