missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human DPPIV/CD26 Quantikine ELISA Kit

Product Code. 18786446
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18786446

Brand: R&D Systems™ DC260B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human DPPIV/CD26 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human DPPIV/CD26 Quantikine ELISA Kit quantifies human DPPIV/CD26 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (10 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.072 ng/mL
Cross Reactivity <0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested
Product Type Human DPPIV/CD26 Quantikine™ ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human DPPIV
For Use With (Application) ELISA
Gene ID (Entrez) 1803
Kit Contents Assay Diluents, Calibrator Diluents, Color Reagent, Conjugate, Microplate, Plate Sealers, Standards, Stop Solution, Wash Buffer
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (10uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2 TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5
Target DPPIV/CD26
Test Time 4.5 hr.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.