missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DRAK1 Synthetic Peptide

Product Code. 15246279
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15246279 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15246279 Supplier Invitrogen™ Supplier No. PEP0051
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 15 amino amino terminus of human DRAK1. PEP-0051 can be used as a blocking peptide with polyclonal antibody PA5-19926.

STK17A (DRAK1) is a serine/threonine kinase localized in the nucleus and involved in apoptosis. Diseases associated with STK17A include Colon Squamous Cell Carcinoma. Among its related pathways are Sweet Taste Signaling. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is STK17B.
TRUSTED_SUSTAINABILITY

Specifica

Gene ID (Entrez) 9263
Species Human
Purification Method purified
Accession Number Q9UEE5
Common Name DRAK1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias DAP kinase-related apoptosis-inducing protein kinase 1; death-associated protein kinase-related 1; DRAK1; serine/threonine kinase 17a; serine/threonine kinase 17a (apoptosis-inducing); serine/threonine-protein kinase 17A; STK17A
Gene Symbol STK17A
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.