missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human EGF Quantikine ELISA Kit

Product Code. p-4753579
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18702004

Brand: R&D Systems™ DEG00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human EGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human EGF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human EGF in cell culture supernates (200 ul), serum (10 ul), edta plasma (200 ul), heparin plasma (200 ul), platelet-poor citrate plasma (200 ul), urine (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.90 to 250 pg/mL
Assay Sensitivity 0.7 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Platelet-poor Plasma (Citrate), Serum, Urine
Specificity Natural and Recombinant Human EGF
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 1950
Gene Symbol EGF
Quantity 1 Plate
Sample Volume Cell Culture Supernates (200uL),Serum (10uL),EDTA Plasma (200uL),Heparin Plasma (200uL),Platelet-poor Citrate Plasma (200uL),Urine (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone
Target EGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.