missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human ENPP-4 (NP_055751) VersaClone cDNA

Product Code. 18793879
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18793879 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18793879 Supplier R&D Systems™ Supplier No. RDC2437
  • ONLINE EXCLUSIVE PRICE
    336.00 €
    317.10 € / 10µg
    Save 18.90 €
    5% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Human ENPP-4 (NP_055751) VersaClone cDNA clone contains the complete ORF for ENPP4, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The ENPP4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Specifications

Gene ENPP4
Accession Number NP_055751
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 22875
Gene Alias AP3Aase, EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative), E-NPP 4, ENPP4, KIAA0879ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function), NPP-4, NPP4ectonucleotide pyrophosphatase/phosphodiesterase family member 4
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.