missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ENTPD3 Partial ORF (NP_001239, 107 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
Description
ENTPD3 is similar to E-type nucleotidases (NTPases). NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD3 contains 4 apyrase-conserved regions which is characteristic of NTPases. [provided by RefSeq]
Sequence: QDVPRAFEECMQKVKGQVPSHLHGSTPIHLGATAGMRLLRLQNETAANEVLESIQSYFKSQPFDFRGAQIISGQEEGVYGWITANYLMGNFLEKNLWHMWVHPHGVETTG
Specifications
Specifications
| Accession Number | NP_001239 |
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 956 |
| Molecular Weight (g/mol) | 37.84kDa |
| Name | ENTPD3 (Human) Recombinant Protein (Q01) |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 ug |
| Immunogen | QDVPRAFEECMQKVKGQVPSHLHGSTPIHLGATAGMRLLRLQNETAANEVLESIQSYFKSQPFDFRGAQIISGQEEGVYGWITANYLMGNFLEKNLWHMWVHPHGVETTG |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction