missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human EphB4 (AAH52804) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
- Also known as: HTK; MYK1; HFASD; CMAVM2; LMPHM7; TYRO11
- EphB4 is a member of the Eph receptor tyrosine kinase family and binds Ephrin-B2.
- Activation of kinase activity occurs after membranebound or clustered ligand recognition and binding.
- EphB4 is expressed preferentially on venous endothelial cells (EC) and inhibits cell-cell adhesion, chemotaxis, and angiogenesis.
- It plays an essential role in vascular development.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hEPHB4 |
| Accession Number | AAH52804 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2977 bp |
| Gene ID (Entrez) | 2050 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction