missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ErbB2 Fc Chimera Recombinant Protein
Click to view available options
Quantity:
5 μg
50 μg
Unit Size:
50µg
5µg
Description
Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
Specifications
Specifications
| Accession Number | P04626 |
| For Use With (Application) | Control, Binding Assay, Bioactivity, Western Blot |
| Formulation | PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4 |
| Gene ID (Entrez) | 2064 |
| Molecular Weight (g/mol) | 96.1 kDa |
| Name | Human ErbB2 Fc Chimera |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | HEK293 cells |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction