missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Factor Va Native Protein

Product Code. 11504161
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11504161 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504161 Supplier Invitrogen™ Supplier No. RP43128
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

RP-43128 contains native human Factor Va protein. RP-43128 may be used as a positive control in many immunological assays. RP-43128 has an Extinction Coefficient (E^1% 1cm, 280 nm) of 17.4. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests, and testing does not always disclose pathogens or other harmful substances contained in blood/plasma products. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12259
Concentration 1.2 mg/mL
For Use With (Application) Bioactivity
Formulation Protein with 5mM CaCl2, 50% glycerol and no preservative
Gene ID (Entrez) 2153
Molecular Weight (g/mol) 168 kDa
Name Human Factor Va
Purification Method SDS-PAGE
Quantity 50 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activated protein C cofactor; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; F5a; factor V; factor V Leiden; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2
Common Name Factor Va
Gene Symbol F5
Biological Activity Specific Activity =2886 U/mg; The specific activity was determined by a Factor V clotting assay, where 1 U (unit) is equivalent to the Factor V activity in 1 mL of normal human plasma.
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Specific Activity =2036 U/mg; The specific activity was determined by a Factor V clotting assay, where 1 U (unit) is equivalent to the Factor V activity in 1 mL of normal human plasma.
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.