missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Factor XII Native Protein

Product Code. 11574111
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11574111 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11574111 Supplier Invitrogen™ Supplier No. RP43076
  • ONLINE EXCLUSIVE PRICE
    177.00 €
    157.53 € / 100µg
    Save 19.47 €
    11% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

RP-43076 contains native human Factor XII protein. RP-43076 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

This single chain zymogen is converted to a two-chain serine protease with an heavy chain (alpha-factor XIIa) and a light chain. The heavy chain contains two fibronectin-type domains, two epidermal growth factor (EGF)-like domains, a kringle domain and a proline-rich domain, whereas the light chain contains only a catalytic domain. On activation, further cleavages takes place in the heavy chain, resulting in the production of beta-factor XIIa light chain and the alpha-factor XIIa light chain becomes beta-factor XIIa heavy chain. Prekallikrein is cleaved by factor XII to form kallikrein, which then cleaves factor XII first to alpha-factor XIIa and then to beta-factor XIIa. The active factor XIIa participates in the initiation of blood coagulation, fibrinolysis, and the generation of bradykinin and angiotensin. It activates coagulation factors VII and XI. Defects in this gene do not cause any clinical symptoms and the sole effect is that whole-blood clotting time is prolonged.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00748
Concentration 2.6 mg/mL
For Use With (Application) Neutralization
Formulation 4mM sodium acetate with 50% glycerol, 150mM NaCl and no preservative; pH 5.3
Gene ID (Entrez) 2161
Molecular Weight (g/mol) 80 kDa
Name Human Factor XII
pH Range 5.3
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias beta-factor XIIa part 1; Beta-factor XIIa part 2; coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; F12; F13; Factor 12; factor XII; Factor12; FXII; HAE3; HAEX; HAF; hageman factor
Common Name Factor XII
Gene Symbol F12
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.