missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Factor XIII Native Protein

Product Code. 11584111
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11584111 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11584111 Supplier Invitrogen™ Supplier No. RP43077
  • ONLINE EXCLUSIVE PRICE
    540.00 €
    480.60 € / 100µg
    Save 59.40 €
    11% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

RP-43077 contains native human Factor XIII protein. RP-43077 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

This gene encodes the coagulation factor XIII A subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon cleavage of the activation peptide by thrombin and in the presence of calcium ion, the plasma factor XIII dissociates its B subunits and yields the same active enzyme, factor XIIIa, as platelet factor XIII. This enzyme acts as a transglutaminase to catalyze the formation of gamma-glutamyl-epsilon-lysine crosslinking between fibrin molecules, thus stabilizing the fibrin clot. It also crosslinks alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin. Factor XIII deficiency is classified into two categories: type I deficiency, characterized by the lack of both the A and B subunits; and type II deficiency, characterized by the lack of the A subunit alone. These defects can result in a lifelong bleeding tendency, defective wound healing, and habitual abortion.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00488
Concentration 6.1 mg/mL
For Use With (Application) Neutralization
Formulation Protein with 0.5M EDTA, 50% glycerol and no preservative
Gene ID (Entrez) 2162
Molecular Weight (g/mol) 320 kDa
Name Human Factor XIII
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias bA525O21.1 (coagulation factor XIII, A1 polypeptide); coagulation factor XIII; coagulation factor XIII A chain; Coagulation factor XIII B chain; coagulation factor XIII, A polypeptide; coagulation factor XIII, A1 polypeptide; coagulation factor XIII, B polypeptide; coagulation factor XIIIa; F13; F13A; F13A1; F13B; Factor 13; factor viiia; factor XIIIa; Factor13; fibrin stabilizing factor, A subunit; fibrinoligase; fibrin-stabilizing factor B subunit; FSF, A subunit; FXIIIB; Protein-glutamine gamma-glutamyltransferase A chain; protein-glutamine gamma-glutamyltransferase B chain; TGase; transglutaminase A chain; Transglutaminase B chain; transglutaminase. plasma
Common Name Factor XIII
Gene Symbol F13A1
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.