missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FBXL20 Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0606

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15378760

  • 130.00 € / 50µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SCRAPPER Synthetic Peptide, PEP-0606, from Invitrogen.

This peptide corresponds to 18 amino acids near the amino terminus of human SCRAPPER.PEP-0606 can be used as a blocking peptide with polyclonal antibody PA5-20486.
Specifications

Specifications

84961
purified
Human FBXL20
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q96IG2
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
FBXL20
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers