missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-8a Recombinant Protein, PeproTech®

Product Code. 17820293
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
Pack of 2
Product Code. Quantity unitSize
17820293 2 x 500 μg 1mg
17810703 100 μg 100µg
30087756 2 x 500 μg Pack of 2
17890493 250 μg 250µg
17810293 25 μg 25µg
17891753 500 μg 500µg
17880493 5 μg 5µg
7 options
This item is not returnable. View return policy

Product Code. 17820293

Brand: Gibco™ 10025A1MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

100-00CC-500UG will be provided as 2 x 250 μg (100-00CC-250UG). CHO cell-derived Recombinant Human FGF-8a is a 21.2 kDa protein consisting of 182 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF-8 is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8a, FGF-8b, FGF-8e and FGF-8f. The human and murine FGF-8a and FGF-8b are identical, unlike human and mouse FGF-8e and FGF-8f, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P55075
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2253
Molecular Weight (g/mol) 21.2
Name Human FGF-8 A
Quantity 2 x 500 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Symbol FGF8
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Sequence QHVREQSLVT DQLSRRLIRT YQLYSRTSGK HVQVLANKRI NAMAEDGDPF AKLIVETDTF GSRVRVRGAE TGLYICMNKK GKLIAKSNGK GKDCVFTEIV LENNYTALQN AKYEGWYMAF TRKGRPRKGS KTRQHQREVH FMKRLPRGHH TTEQSLRFEF LNYPPFTRSL RGSQRTWAPE PR
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.