missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FKBP12 (aa 1-13) Synthetic Peptide

Product Code. 11554670
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11554670 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11554670 Supplier Invitrogen™ Supplier No. PEP012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Human FKBP12 (residues1-13) Synthetic Peptide, PEP-012, from Invitrogen.

PEP-012 is a 14 amino acid (amino acids 1-13) synthetic peptide whose sequence is from the amino end of the human FKBP12 protein. The sequence of this peptide is (amino to carboxy terminus): G1-V-Q-V-E-T-I-S-P-G-D-G-R13-C (The C-terminal cysteine has been added to facilitate conjugation and is not a part of the native human FKBP12 sequence)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and FKBP12, catalog # PA1-026A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2280
Species Human
Purification Method purified
Accession Number P62942
Common Name Human FKBP12 (aa 1-13)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol FKBP1A
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence G1 - V - Q - V - E - T - I - S - P - G - D - G - R13 - C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.