missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FNDC1 (aa 1535-1633) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56603 (PA5-56603. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q4ZHG4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 84624 |
| Name | Human FNDC1 (aa 1535-1633) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1110027O12Rik; 1110051A18Rik; Activation-associated cDNA protein; activator of G-protein signaling 8; AGS8; bA243O10.1; dJ322A24.1; expressed in synovial lining protein; fibronectin type III domain containing 1; fibronectin type III domain containing 2; fibronectin type III domain-containing protein 1; FNDC1; FNDC2; KIAA1866; MEL4B3; mKIAA1866 |
| Common Name | FNDC1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction