missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Human, Forward Primer, Desalted

Product Code. 15574364 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
25 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15574364 25 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574364 Supplier Applied Biosystems™ Supplier No. A15629
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human, Forward Primer, Desalted

Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.

TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Single Tube, Dried-down
Format Solid
For Use With (Equipment) Mass Spectrometry
Description Forward PCR/SEQ Primer, 25 nmole, Solid, Desalted Purification, Hs00196555_CE Primer ID, 8nmole Minimal Spec Yield, Non-tailed Primer 5'-tail Option, Human Species, Single Tube, Delivered in a Single tube, Dried-down, Used for PCR and Cycle Sequencing Reactions
Product Type Forward PCR or SEQ Primer
Selection Agent (Eukaryotic) Human
Purification Method Desalted
Purity Desalted
Primer Forward PCR/SEQ
Quantity 25 nmol
Grade HPLC
For Use With (Application) PCR and Cycle Sequencing Reactions
Form Solid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.