Learn More
Abnova™ Human FRAT1 Partial ORF (NP_005470.2, 181 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010023-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene belongs to the GSK-3-binding protein family. The protein inhibits GSK-3-mediated phosphorylation of beta-catenin and positively regulates the Wnt signaling pathway. It may function in tumor progression and in lymphomagenesis. [provided by RefSeq]
Sequence: RLQQRRGSQPETRTGDDDPHRLLQQLVLSGNLIKEAVRRLHSRRLQLRAKLPQRPLLGPLSAPVHEPPSPRSPRAACSDPGASGRAQLRTGDGVLVPGSSpecifications
NP_005470.2 | |
Liquid | |
10023 | |
FRAT1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ97193 | |
FRAT1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RLQQRRGSQPETRTGDDDPHRLLQQLVLSGNLIKEAVRRLHSRRLQLRAKLPQRPLLGPLSAPVHEPPSPRSPRAACSDPGASGRAQLRTGDGVLVPGS | |
RUO | |
FRAT1 | |
Wheat Germ (in vitro) | |
GST |