missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Fructosamine-3-kinase (NP_071441.1) VersaClone cDNA

Product Code. 18722337
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722337 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722337 Supplier R&D Systems™ Supplier No. RDC3777
  • ONLINE EXCLUSIVE PRICE
    365.00 €
    344.40 € / 10µg
    Save 20.60 €
    5% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Gene FN3K
Accession Number NP_071441.1
Gene ID (Entrez) 64122
Gene Alias EC 2.7.1.-, FN3K, fructosamine 3 kinase, fructosamine-3-kinase
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.