missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Gamma Globulin

Product Code. 11859220
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11859220 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11859220 Supplier Invitrogen™ Supplier No. 31879
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Storage: After opening, dilute only enough antibody for a single day's use. Country of Origin: USA.

Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Liquid
Product Type Human Gamma Globulin
Quantity 10 mg
Concentration 11.9 mg/mL
Formulation PBS with no preservative; pH 7.6
For Use With (Application) Control, ChIP Assay, Immunoprecipitation, Flow Cytometry
Isotype Ig
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.