missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human GDF-15/MIC-1 Recombinant Protein, PeproTech®

Product Code. p-7246026
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17810303

Brand: Gibco™ 12028C100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human GDF-15/MIC-1 is a disulfide linked homodimeric protein consisting of two 112 amino acid polypeptide chains. The calculated molecular weight of Recombinant Human GDF-15/MIC-1 is 24.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDF-15 belongs to the TGF-beta cytokine family, whose members play an important role during prenatal development and postnatal growth, and the remodeling and maintenance of a variety of tissues and organs. GDF-15 is expressed predominantly in the placenta and, to a much lesser extent, in various other tissues. The presence of GDF-15 in amniotic fluid and its elevated levels in the sera of pregnant women suggest GDF-15's involvement in gestation and embryonic development. GDF-15 generally exerts tumor suppressive activities and is one of the predominant factors produced and secreted in response to activation of the p53 pathway. Interestingly, the serum level of GDF-15 is positively correlated with neoplastic progression of several tumor types, including certain colorectal, pancreatic, and prostate cancers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99988
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 9518
Molecular Weight (g/mol) 24.6 kDa
Name Human GDF-15/MIC-1
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Gdf15; GDF-15; growth differentiation factor 15; growth/differentiation factor 15; macrophage inhibiting cytokine-1; macrophage inhibitory cytokine 1; MIC1; MIC-1; NAG1; NAG-1; NRG-1; NSAID (nonsteroidal anti-inflammatory drug)-activated protein 1; NSAID-activated gene 1 protein; NSAID-regulated gene 1 protein; PDF; PLAB; Placental bone morphogenetic protein; placental TGF-beta; Prostate differentiation factor; PTGFB; PTGF-beta; SBF
Common Name GDF15
Gene Symbol Gdf15
Biological Activity Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1 osteoblastcells. The expected ED50 for this effect is 75-200 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence ARNGDHCPLG PGRCCRLHTV RASLEDLGWA DWVLSPREVQ VTMCIGACPS QFRAANMHAQ IKTSLHRLKP DTVPAPCCVP ASYNPMVLIQ KTDTGVSLQT YDDLLAKDCH CI
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.