Learn More
Invitrogen™ Human Glucocorticoid Receptor (aa 245-259) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP002
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human Glucocorticoid Receptor (residues 245-259) Synthetic Peptide, PEP-002, from Invitrogen.
PEP-002 is a 15 amino acid (amino acids 245-259) synthetic peptide whose sequence is from the amino terminus of the human glucocorticoid receptor (GR). The sequence of this peptide is (amino to carboxy terminus): C-K-P-L-I-L-P-D-T-K-P-K-I-K-D (The N-terminal cysteine has been added to facilitate conjugation and is not a part of the native human GR sequence)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and the human GR, catalog # PA1-512. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Specifications
2908 | |
purified | |
Human Glucocorticoid Receptor (aa 245-259) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P04150 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
NR3C1 | |
No preservative | |
50 μg | |
C - K - P - L - I - L - P - D - T - K - P - K - I - K - D |