missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Hepcidin Quantikine ELISA Kit

Product Code. 18791864
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18791864 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18791864 Supplier R&D Systems™ Supplier No. DHP250
  • ONLINE EXCLUSIVE PRICE
    893.00 €
    844.20 € / 1 sets
    Save 48.80 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Hepcidin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Hepcidin Quantikine ELISA Kit quantifies human Hepcidin Antimicrobial Peptide in cell culture supernates (25 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity 3.81pg/mL
Product Line R&D Systems
Sample Type Serum, Urine
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.