missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Human HPRT1 (HGPRT) Endogenous Control (FAM™/MGB probe, non-primer limited)

Product Code. 10534255 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
125 Reactions
500 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10534255 125 Reactions Each
10724037 500 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10534255 Supplier Applied Biosystems™ Supplier No. 4333768T
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human HPRT1 (HGPRT) Endogenous Control (FAM™/MGB probe, non-primer limited)

The Applied Biosystems™ Human HPRT1 (HGPRT) Endogenous Control (FAM™ ⁄ MGB Probe, Non-Primer Limited) is intended as an endogenous control. It allows relative gene expression quantification in cDNA samples when used with other gene expression assays. Probe is labeled with 6FAM™ dye - MGB and the primers are not limited. Can be used for singleplex PCR reactions. Endogenous control is to be used with Inventoried and Non-Inventoried TaqMan™ Gene Expression Assays, Custom TaqMan™ Gene Expression Assays, and⁄or Custom TaqMan™ Primers and Probes.

Assay Details:

Gene Symbol: HPRT1
RefSeq: NM_000194.1
Probe Exon Location:6-7
Amplicon Size: 100
Corresponding TaqMan Assay ID: Hs99999909_m1

TaqMan™ Endogenous Controls

Eliminate months of assay design, formulation, and testing by using TaqMan™ Endogenous Controls as your controls to quantify gene expression. This convenient collection of pre-designed probe and primer sets enables you to normalize the amount of sample RNA or DNA in a reaction.

Complete Solution for Quantitative Gene Expression

Having a hard time deciding what controls to use to quantify gene expression — even with detailed information on biological systems? Now, with TaqMan™ Endogenous Controls, you can avoid all the trial-and-error of selecting controls for most common human, mouse, rat, and eukaryotic genes.

Simple to Use

All components of the TaqMan™ Endogenous Controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan™ Universal PCR Master Mix (with or without AmpErase™ UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.

Flexible Offering

We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
  • A FAM™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (900nM each)
  • A VIC™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)
  • A VIC™ dye-labeled TAMRA™ probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)

    Choosing the Right Endogenous Control

    Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.

    Multiplex vs. Singleplex PCR

    All TaqMan™ Endogenous Controls that contain probes labeled with the VIC™ reporter dye are primer limited. This allows multiplexing of TaqMan™ Endogenous Controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan™ Endogenous Controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing .

    For Research Use Only. Not for use in diagnostic procedures.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Control primer-probe sets, 1 tube containing 20X mix of pre-formulated assay (1 probe and 2 primers)
    Store at –15 to –25°C
    Description Human HPRT1 (HGPRT) Endogenous Control, 125rxns, Liquid, Tube(s) Format, FAM™ Dye Type, HPRT1 Target Gene, Human Species, Format, –15 to –25°C Recommended Storage, For Real-Time PCR (qPCR), Real-Time PCR-Based Gene Expression Profiling
    Detection Method Primer-probe
    PCR Method 2-step RT-qPCR
    Quantity 125 rxn(s)
    For Use With (Application) Gene Expression
    No. of Reactions 125 Reactions

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.