missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IFN-alpha 2 (alpha 2b) Recombinant Protein

Recombinant Protein

Brand:  Invitrogen™ 111051

Product Code. 10005793

  • 328.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

After receipt, the product may be stored at -20°C for short-term use (less than 6 months). For long-term storage, store the product at -70°C or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed. Aliquot and refreeze unused contents to minimize effects of freeze thawing. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of product should be in buffers containing protein such as 0.1% BSA or tissue culture media with serum.

Interferon alpha-2b is a cytokine that belongs to the type I interferon family and is encoded by a gene cluster on chromosome 9. It is produced in response to viral infection and plays a key role in the innate immune response, exhibiting potent antiviral, antiproliferative and immunomodulatory properties. Interferon alpha-2b binds to a plasma membrane receptor consisting of IFNAR1 and IFNAR2, which is expressed ubiquitously in the body and can thus act on virtually all cells. This cytokine is effective in reducing the symptoms and duration of the common cold, and has shown efficacy in treating various types of cancer, including hematological malignancies and solid tumors. A deficiency of type I interferon in the blood is considered a hallmark of severe COVID-19, and a combination therapy involving interferon alpha-2b may be a potential therapeutic approach. IFNA2 has been associated with diseases such as Severe Covid-19 and Hepatitis B.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P01563
ELISA
3440
Human IFN-alpha 2 (alpha 2 b)
E. Coli
<1 EU/ μg
Interferon alpha-2 b
EC50 ̃ 1 U/mL; determined by use of the cytopathic effect inhibition assay described in [Rubinstein, S. et al. (1981). J. Virol. 37, 755-758 ]; [Familletti, P.C. et al. (1981). Methods in Enzymology, Vol. 78, 387-394 ]. The units are determined with respect to the international reference standard for human interferon alpha A (Hu-IFN-alpha-A) provided by the National Institutes of Health [see Pestka, S. (1986). Methods in Enzymology, Academic Press, New York 119, 14-23 ]. Unit of activity measured on bovine MDBK cells with vesicular stomatitis virus (VSV).
E. coli
-80°C, Avoid Freeze/Thaw Cycles
Liquid
>95%
Lot-specific
PBS with 0.1% BSA and no preservative
19.3 kDa
1 x 10^5 units
RUO
alpha-2 A interferon; IFNA; IFNA2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha-2; Interferon alpha-A; interferon, alpha 2; leIF A
IFNA2
Unconjugated
Recombinant
E. coli
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.