missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IGF-II/IGF2 Quantikine ELISA Kit

Product Code. 30000837
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30000837

Brand: R&D Systems™ DG200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

E. coli-expressed recombinant human IGF-II

Human IGF-II/IGF2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IGF-II/IGF2 Quantikine ELISA Kit quantifies human IGF-II/IGF2 in serum-free cell culture supernates (25 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Description C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PP9974, somatomedin-A
Product Type ELISA Kit
Quantity 1 Kit
Species Human
Assay Sensitivity 5.41 pg/mL
Assay Range 12.50 to 800 pg/mL
Cross Reactivity <0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested
For Use With (Application) ELISA
Gene ID (Entrez) 3481
Sample Type Serum-free Cell Culture Supernates (25uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Urine (25uL)
Sample Volume 25 μL (Serum-free Cell Culture Supernates), 10 μL (Serum), EDTA
Test Time 3.5 hr.
Synonym C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A
Kit Contents E. coli-expressed recombinant human IGF-II
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.