missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-10 Quantikine ELISA Kit

Product Code. 18752064
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752064 1 Plate 1 set
18792294 1 Pk. (6 Plates) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752064 Supplier R&D Systems™ Supplier No. D1000B
  • ONLINE EXCLUSIVE PRICE
    854.30 €
    760.20 € / 1 sets
    Save 94.10 €
    11% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-10 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-10 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-10 in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul), citrate plasma (200 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.80 to 500 pg/mL
Assay Sensitivity 3.9 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Citrate, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human IL-10
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3586
Gene Symbol IL10
Quantity 1 Plate
Sample Volume Cell Culture Supernates (200 μL), Serum (200 μL), EDTA Plasma (200 μL), Heparin Plasma (200 μL), Citrate Plasma (200 μL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF
Target IL-10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.