Learn More
R&D Systems™ Human IL-11 (NP_000632.1) VersaClone cDNA
Product Code. 18781777
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity Unit Size
18781777 10 μg 10µg
1 options
Product Code. 18781777 Supplier R&D Systems™ Supplier No. RDC3696
  • ONLINE EXCLUSIVE PRICE
    304.89 €
    270.90 € / 10µg
    Save 33.99 €
    11% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Gene IL-11
Accession Number NP_000632.1
Gene ID (Entrez) 3589
Gene Alias Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11 Oprelvekin, interleukin 11, interleukin-11
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems