missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Recognizes human IL13
- IL13 (Interleukin 13) is an immunoregulatory cytokine produced primarily by activated Th2 cells
- Cytokine involved in stages of B-cell maturation and differentiation
- Regulates CD23 and MHC class II expression and promotes IgE isotype switching of B cells
- Inhibits production of series of cytokines by activated human monocytes
- Induces IFN-γ production by NK cells
- Thought to be important cytokine in pathogenesis of asthma and shows pivotal role in fibrotic diseases (hepatic and pulmonary fibrosis, nodular sclerosing Hodgkin's disease)
Process:
- Antibody specific for IL13 pre-coated onto microwells
- IL13 protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody of biotinylated specific for IL13 is added to detect captured IL13 protein
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | NP_002179 |
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 1 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 3596 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Human IL-13 ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?