missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-13 Recombinant Protein

Product Code. 17965821
Click to view available options
Quantity:
1 mg
100 μg
5 μg
Unit Size:
100µg
1mg
5µg
This item is not returnable. View return policy

Product Code. 17965821

Brand: Invitrogen™ RP88069

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. Reconstitute with sterile deionized water to 0.1 mg/mL. Concentration is measured by UV.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35225
Concentration 0.25 mg/mL
For Use With (Application) Control, Bioactivity
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 3596
Molecular Weight (g/mol) 12.35 kDa
Name Human IL-13
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <0.01 EU/μg
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Common Name IL-13
Gene Symbol Il13
Biological Activity Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323.). The ED50 for this effect is typically 0.5-3 ng/mL
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Human IL13, amino acids Met1-Asn132 (Accession # AAK53823.1)
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured in a cell proliferation assay using TF1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323.). The ED50 for this effect is typically 0.5-3 ng/mL.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥95% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.