missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-17A Recombinant Protein

Product Code. 10029132
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10029132

Brand: Gibco™ PHC9171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Lyophilized human IL-17A should be reconstituted in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. For maximal stability, a carrier protein, such as 0.1-1.0% BSA, may be included. The reconstituted protein should be apportioned into working aliquots and stored at ≤-20°C. Further dilutions should be made in low endotoxin medium or buffered solution with FBS or tissue culture grade BSA. Storage: For maximal stability, store this lyophilized preparation at ≤-20°C, preferably desiccated. For shipment and short term storage (up to 2 weeks), storage at 2°C to 8°C is sufficient. Reconstituted human IL-17A should be stored at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16552
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3605
Molecular Weight (g/mol) 15.5 kDa
Name Human IL-17A
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A
Common Name IL-17A
Gene Symbol IL17A
Biological Activity ED50 = 0.5 - 5.0 ng/mL; determined by the dose-dependent production of GRO-alpha by HT-29 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Human IL-17A recombinant protein is homodimeric containing two 137 amino acid subunits
Content And Storage -20°C
Classification Carrier-Free
Activity ED50 = 0.5 - 5.0 ng/mL; determined by the dose-dependent production of GRO-alpha by HT-29 cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.