missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-1b High Sensitivity ELISA Kit

Product Code. 30054496
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30054496 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30054496 Supplier Invitrogen™ Supplier No. BMS2242HS
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

ELISA

The Human High Sensitivity Interleukin 1 beta (Hu HS IL-1B) ELISA quantitates Hu HS IL-1β in human serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu HS IL-1beta Principle of the method The Human HS IL-1B solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A special amplification reagent step is added to achieve the high sensitivity capability. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01584
Assay Range 0,16 to 10,0
Assay Sensitivity 0.05 pg/mL
Description Human IL-1b High Sensitivity ELISA Kit
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3553
Gene Symbol IL1B
Interassay CV 8.1%
Intraassay CV 6.7
Kit Contents Pre-coated microwell plate, Biotinylated Detection Antibody, Streptavidin-HRP, Standard, Sample Diluent, Assay Buffer Concentrate, Amplification Reagent I, Amplification Reagent II, Wash Buffer Concentrate, Substrate Solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 50 μL
Storage Requirements Store kit at 2°C to 8°C. Refer to product documentation for component specific storage temperature.
Target IL-1 beta
Test Time 1 hrs 20 mins
Total Assay Time 18 hrs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.