missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-2 Quantikine HS ELISA Kit

Product Code. 18740977
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
18740977 Store the unopened product at 2°C to 8°C. Do not use past expiration date. 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18740977 Supplier R&D Systems™ Supplier No. HS200
  • ONLINE EXCLUSIVE PRICE
    1022.09 €
    912.45 € / 1 sets
    Save 109.64 €
    10% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-2 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-2 Quantikine HS ELISA Kit quantifies human IL-2 in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Description 0.066pg/mL sensitivity
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Type Human IL-2 Quantikine HS ELISA Kit
Gene ID (Entrez) 3558 (Human); 16183 (Mouse); 116562 (Rat); 396868 (Porcine); 280822 (Bovine); 403989 (Canine); 100034204 (Equine); 751114 (Feline); 100302458 (Rabbit)
Includes Microplate, conjugate, standards, assay diluents, calibrator diluents, wash buffer, color reagent, stop Solution, plate sealers
Intended Use Measure human IL-2 levels in serum and plasma
Product Line R&D Systems
Time Range 4 hours

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.