missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-33 DuoSet ELISA

Product Code. 30021023
Change view
Click to view available options
Quantity:
1 Kit (for 15 Plates)
1 Kit (for 5 Plates)
Unit Size:
1 set
Pack of 15
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30021023 1 Kit (for 5 Plates) 1 set
30020997 1 Kit (for 15 Plates) 1 set
30000772 1 Kit (for 15 Plates) Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30021023 Supplier R&D Systems™ Supplier No. DY3625B05
  • ONLINE EXCLUSIVE PRICE
    397.99 €
    354.90 € / 1 sets
    Save 43.09 €
    10% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

Human IL-33 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-33 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-33.
TRUSTED_SUSTAINABILITY

Specifications

Description Fucose Detection
Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Assay Range 23.40 to 1500 pg/mL
Cross Reactivity Not observed
Format 96-well Strip Plate
For Use With (Application) ELISA
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Serum, Plasma
Kit Contents Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cytokines
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.