missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-34 Quantikine ELISA Kit

Product Code. 18752114
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
18752114 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18752114 Supplier R&D Systems™ Supplier No. D3400
  • ONLINE EXCLUSIVE PRICE
    946.38 €
    844.20 € / 1 sets
    Save 102.18 €
    10% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-34 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-34 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-34 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (50 ul), human milk (13 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.30 - 2000pg/mL
Assay Sensitivity 3.06pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum
Specificity Natural and Recombinant Human IL-34
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 146433
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (50uL),Human Milk (13uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C16orf77, IL34, IL-34, interleukin 34
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.