missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-34 Recombinant Protein, PeproTech®

Product Code. 17870433
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17870433 250 μg 250µg
17821883 100 μg 100µg
17850433 10 μg 10µg
30089736 2 x 500 μg Pack of 2
17810063 2 x 500 μg 1mg
17821813 2 μg 2µg
17850443 500 μg 500µg
17871873 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17870433 Supplier Gibco™ Supplier No. 20034250UG

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Recombinant Protein

200-34-1MG will be provided as 2 x 500 μg (200-34-500UG). Recombinant Human IL-34 is a 52.5 kDa homodimeric glycoprotein consisting of 460 amino acid residues, including a C-terminal His-Tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6ZMJ4
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 146433
Molecular Weight (g/mol) 52.5 kDa
Name Human IL-34
Purification Method Purified
Quantity 250 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34
Common Name IL-34
Gene Symbol Il34
Biological Activity Measured by its ability to induce secretion of MCP-1 from PBMCs.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP KVESVLSLLN APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHHHH
Content And Storage -20°C
Activity Measured by its ability to induce secretion of MCP-1 from PBMCs.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.