missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-4 Recombinant Protein, PeproTech®
Description
Recombinant Human IL-4 is a 15.1 kDa globular protein containing 130 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Specifications
Specifications
| Accession Number | P05112 |
| For Use With (Application) | Control, Western Blot Control, ELISA Standard, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3565 |
| Molecular Weight (g/mol) | 15.1 kDa |
| Name | Human IL-4 |
| Purification Method | Purified |
| Quantity | 100 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?