missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-6 DuoSet ELISA

Product Code. 30021411
Change view
Click to view available options
Packaging Quantity:
15 Plates
5 Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
30021411 15 Plates 1 set
30021312 5 Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30021411 Supplier R&D Systems™ Supplier No. DY206
  • ONLINE EXCLUSIVE PRICE
    777.56 €
    693.00 € / 1 sets
    Save 84.56 €
    10% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-6 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-6 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-6.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit
Quantity 1 Kit (for 15 Plates)
Species Human
Conjugate Biotin
Assay Range 9.38 to 600 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Product Line R&D Systems
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cytokines
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.