missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-7 Recombinant Protein, PeproTech®
Description
200-07-1MG will be provided as 2 x 500 μg (200-07-500UG). Recombinant Human IL-7 is a 17.5 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications
Specifications
| Accession Number | P13232, P16871 |
| For Use With (Application) | Bioactivity, Control, ELISA Standard, Western Blot Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3574, 3575 |
| Molecular Weight (g/mol) | 17.5 kDa |
| Name | Human IL-7 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?