missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-8/CXCL8 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
1 Kit (for 5 Plates)
Unit Size:
1 set
Description
Requires
DuoSet Ancillary Reagent Kit 2 - Catalog # DY008
OR
- Wash Buffer - 0.05%Tween™ 20 in PBS, pH 7.2-7.4
- Block Buffer - 1% BSA in PBS, pH 7.2-7.4
- Reagent Diluent - 0.1% BSA, 0.05% Tween 20 in Tris-buffered Saline, pH 7.2-7.4, 0.2μm filtered
- Substrate Solution - Catalog # DY999
- Stop Solution - 2N H2SO4
- Microplates - Catalog # DY990
- ELISA Plate Sealers
The Human IL-8/CXCL8 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CXCL8/IL-8.
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA Development Kit |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Sensitivity | 31.20 to 2000 pg/mL |
| Assay Range | 31.20 to 2000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3576 |
| Synonym | 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction