missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-8/CXCL8 Quantikine ELISA Kit

Product Code. 18792354
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sufficient For Unit Size
18792354 1 Plate 1 x 96-well Plate 1 set
18712314 6 Plates 6 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792354 Supplier R&D Systems™ Supplier No. D8000C
  • ONLINE EXCLUSIVE PRICE
    854.30 €
    760.20 € / 1 sets
    Save 94.10 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human IL-8/CXCL8 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-8/CXCL8 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL8/IL-8 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 7.5pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human IL-8
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50 μL), Serum (50 μL), EDTA Plasma (50 μL), Heparin Plasma (50 μL), Citrate Plasma (50 μL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym 3-10 C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1 TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1 NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10 C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.