missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Layilin (NP_001245319) VersaClone cDNA
Description
- Layilin (named after the L-A-Y-I-L-I six amino acid motif in the molecule’s transmembrane segment) is a member of the animal C-type lectin family.
- Layilin is known to be a receptor for hyaluronan (HA), a negatively charged linear high molecular weight polysaccharide composed of alternating units of D-glucuronic acid and N-acetyl-D-glucosamine.
- HA is found in the extracellular matrix of most animal tissues and in body fluids.
- Thus, it may be that layilin modulates cell behavior and functions during tissue remodeling, development, homeostasis, and disease.
- Layilin binds specifically to HA but not to other glycosaminoglycans.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hLAYN |
| Accession Number | NP_001245319 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1162 bp |
| Gene ID (Entrez) | 143903 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?