missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LBP ELISA Kit

Product Code. 17134593
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17134593 96 Tests Pack of 96
17115069 5 x 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17134593 Supplier Invitrogen™ Supplier No. EH297RB
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

ELISA

Human LBP quantitates human LBP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LBP.

The LBP protein is involved in the acute-phase immunologic response to gram-negative bacterial infections. Gram-negative bacteria contain a glycolipid, lipopolysaccharide (LPS), on their outer cell wall. Together with bactericidal permeability-increasing protein (BPI), the encoded protein binds LPS and interacts with the CD14 receptor, probably playing a role in regulating LPS-dependent monocyte responses. This protein is part of a family of structurally and functionally related proteins, including BPI, plasma cholesteryl ester transfer protein (CETP), and phospholipid transfer protein (PLTP). Finally, this gene is found on chromosome 20, immediately downstream of the BPI gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18428
Assay Range 0.82-200 ng/mL
Assay Sensitivity 0.82 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias acute phase protein; binds bacterial LPS; BPI fold containing family D, member 2; Bpifd2; hypothetical protein LOC512242; LBP; lipopolysaccharide binding protein; lipopolysaccharide-binding protein; lipopolysaccharide-binding protein LPSBP; LPS-binding protein; LPSBP; Ly88acute phase protein; binds bacterial LPS; BPI fold containing family D, member 2; Bpifd2; hypothetical protein LOC512242; LBP; lipopolysaccharide binding protein; lipopolysaccharide-binding protein; lipopolysaccharide-binding protein LPSBP; LPS-binding protein; LPSBP; Ly88;
Gene ID (Entrez) 3929
Gene Symbol LBP
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target LBP
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.