missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LGI2 Synthetic Peptide

Product Code. 15358770
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15358770 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15358770 Supplier Invitrogen™ Supplier No. PEP0613
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 10 amino acids near the center of human LGI2. PEP-0613 can be used as a blocking peptide with polyclonal antibody PA5-20493.

The leucine-rich, glioma inactivated gene 2 (LGI2) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI2 have not been found to be associated with ADTLE.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 55203
Species Human
Purification Method purified
Accession Number Q8N0V4
Common Name LGI2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias Kiaa1916; leucine rich repeat LGI family member 2; leucine-rich glioma-inactivated protein 2; Leucine-rich repeat LGI family member 2; leucine-rich repeat LGI family, member 2; leucine-rich, glioma inactivated 2; LGI1-like protein 2; Lgi2; LGIL2; mKIAA1916
Gene Symbol Lgi2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.